missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V delta 2 Antibody (B6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11706F
This item is not returnable.
View return policy
Description
TCR V delta 2 Monoclonal antibody specifically detects TCR V delta 2 in Human samples. It is validated for Flow Cytometry
Specifications
| TCR V delta 2 | |
| Monoclonal | |
| FITC | |
| hDV102S1 | |
| Human recombinant TCR Vdelta2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| B6 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 28517 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction