missing translation for 'onlineSavingsMsg'
Learn More

Glut2 Antibody (37E1), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 343.00 - € 533.00

Specifications

Antigen Glut2
Clone 37E1
Dilution Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30038602
View Documents
Novus Biologicals
NBP3-26318-50ul
50 μL
€ 343.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30031683
View Documents
Novus Biologicals
NBP3-26318-100ul
100 μL
€ 533.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glut2 Monoclonal antibody specifically detects Glut2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Specifications

Specifications

Glut2
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human
GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2
A synthesized peptide derived from Human Glut2 [UniProt P11168]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
37E1
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Unconjugated
Rabbit
Lipid and Metabolism, Stem Cell Signaling Pathway
PBS, pH 7.4, 150mM NaCl and 50% glycerol
6514
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.