missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20436-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detected 60-70 kDa fully glycosylated, matured GLUT2 and a nonglycosylated form of GLUT2 around 38-45 kDa.
Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.Specifications
| GLUT2 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A2 | |
| GLUT 2, GLUT2, SLC2A2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20526, 25351, 6514 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11168, P12336, P14246 | |
| Slc2a2 | |
| GLUT2 Fusion Protein Ag14204 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction