missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLUT2 Recombinant Rabbit Monoclonal Antibody (37E1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550125
This item is not returnable.
View return policy
Description
GLUT2 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Flow, ELISA
Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.
Specifications
| GLUT2 | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11168 | |
| Slc2a2 | |
| A synthesized peptide derived from Human SLC2A2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 37E1 | |
| Unconjugated | |
| Slc2a2 | |
| AI266973; glucose transporter 2; glucose transporter type 2, liver; GLUT2; GLUT-2; GTT2; liver-type glucose transporter; Slc2a2; solute carrier family 2 (facilitated glucose transporter), member 2; solute carrier family 2 member 2; solute carrier family 2, facilitated glucose transporter member 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6514 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction