missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut3 Antibody (202017), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1415I
This item is not returnable.
View return policy
Description
Glut3 Monoclonal antibody specifically detects Glut3 in Human samples. It is validated for Flow Cytometry
Specifications
| Glut3 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| FLJ90380, GLUT-3, GLUT3Glucose transporter type 3, brain, solute carrier family 2 (facilitated glucose transporter), member 3, solute carrier family 2, facilitated glucose transporter member 3 | |
| NS0 mouse myeloma cell line transfected with human Glut3, Accession # P11169 | |
| 0.2 mL | |
| Alzheimers Research, Cancer, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 6515 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 202017 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction