missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLUT3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596952

Product Code. 16599781

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ESPRFLLINR KEEENAKQIL QRLWGTQDVS QDIQEMKDES ARMSQEKQVT VLELFRVSSY RQPIIISIVL QLSQQ Positive Samples: HeLa, HepG2, U-87MG; Cellular Location: Cell membrane, Multi-pass membrane protein.

Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF1 induced signaling. GLUT3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT3 staining is also detectable in human cancers including those of the ovary, lung, and testis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GLUT3
Polyclonal
Unconjugated
SLC2A3
AA408729; AL023014; AL024341; AU040424; C78366; FLJ90380; glucose transporter 3; glucose transporter type 3, brain; Glut3; Glut-3; Slc2a3; solute carrier family 2 (facilitated glucose transporter), member 3; Solute carrier family 2 A3 (neuron glucose transporter); solute carrier family 2 member 3; solute carrier family 2, facilitated glucose transporter member 3; solute carrier family 2, member 2; solute carrier family 2, member 3
Rabbit
Affinity Chromatography
RUO
20527, 25551, 6515
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1.09 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
P11169, P32037, Q07647
SLC2A3
Recombinant fusion protein containing a sequence corresponding to amino acids 207-281 of human GLUT3/SLC2A3 (NP_0088621).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.