missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut4 Antibody (6H11NB), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44298V
This item is not returnable.
View return policy
Description
Glut4 Monoclonal antibody specifically detects Glut4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Glut4 | |
| Monoclonal | |
| DyLight 405 | |
| GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4 | |
| Partial recombinant human Glut4 protein (between amino acids 300-350) [Uniprot: P14672] | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroendocrinology | |
| 6517 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6H11NB | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction