missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut5 Antibody (OTI9F3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71285FR
This item is not returnable.
View return policy
Description
Glut5 Monoclonal specifically detects Glut5 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Glut5 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| SLC2A5 | |
| Full length recombinant protein of human Glut5 (NP_003030) produced in HEK293T cells. | |
| 0.1 mL | |
| 6518.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI9F3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Fructose transporter, Glucose transporter type 5, small intestine, GLUT-5, GLUT5glucose transporter-like protein 5, solute carrier family 2 (facilitated glucose transporter), member 5, solute carrier family 2 (facilitated glucose/fructose transporter), member 5, solute carrier family 2, facilitated glucose transporter member 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction