missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glutamate Receptor 6 Antibody (818235) [PE/Cy5.5], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ FAB9610PECY55
This item is not returnable.
View return policy
Description
Glutamate Receptor 6 Monoclonal antibody specifically detects Glutamate Receptor 6 in Human samples. It is validated for Flow Cytometry
Specifications
| Glutamate Receptor 6 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427 | |
| Chinese Hamster Ovary cell line CHO-derived human GluR6/GRIK2 protein, Thr32 - Asn549, Accession # Q13002 | |
| 0.1 mL | |
| Apoptosis, Cell Biology, GPCR, Neuroscience, Signal Transduction | |
| 2898 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 818235 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction