missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glutaminase C (GAC) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540134
This item is not returnable.
View return policy
Description
This antibody specifically recognizes GAC but not the KGA isoform. Predicted reactivity: Bovine (100%). Recommended positive controls: MDA-MB-231 and cholangiocarcinoma tissue. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Sahai (1983) demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter(Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.
Specifications
| Glutaminase C (GAC) | |
| Polyclonal | |
| Unconjugated | |
| GLS | |
| 6330442B14; AAD20; AI314027; B230365M23Rik; GAC; GAM; GLS; Gls1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; RATGLUT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14660, 2744 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| D3Z7P3, O94925 | |
| GLS | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Glutaminase C (GAC). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction