Learn More
Glutamine Synthetase Antibody (OTI1F4), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-70834IR
Product Code. 18155934
  • ONLINE EXCLUSIVE PRICE
    € 785.00
    € 742.35 / 0.10mL
    Save € 42.65
    5% Off
Description
Glutamine Synthetase Monoclonal specifically detects Glutamine Synthetase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glutamine Synthetase
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
GLUL
Full length recombinant protein of human Glutamine Synthetase (NP_002056) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1F4
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Mouse
Affinity Purified
Hypoxia, Neurodegeneration, Neuroscience, Vision
2752.0
Store at 4°C in the dark.