missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine Synthetase Antibody (rGLUL/8621), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24286AF405
This item is not returnable.
View return policy
Description
Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Glutamine Synthetase | |
| Monoclonal | |
| Alexa Fluor 405 | |
| cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43 | |
| Recombinant fragment (around aa 1-200) of human Glutamine Synthetase protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Hypoxia, Neurodegeneration, Neuroscience, Vision | |
| 2752 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rGLUL/8621 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction