Learn More
Glutaminyl-peptide Cyclotransferase/QPCT Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-00186PCP
Product Code. 18659086
  • € 430.00 / 0.10mL
Description
Glutaminyl-peptide Cyclotransferase/QPCT Polyclonal antibody specifically detects Glutaminyl-peptide Cyclotransferase/QPCT in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PBS
Antigen and protein A Affinity-purified
0.1 mL
25797