missing translation for 'onlineSavingsMsg'
Learn More

Glutathione (glycine-13C2,15N) Sodium, TRC

Product Code. 30012704
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012704 1 mg 1mg
30010664 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012704 Supplier Toronto Research Chemicals Supplier No. TRCG5979521MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C813C2H17N215NO6S •xNa
Molecular Weight (g/mol) 310.302299
Synonym L-Glutamyl-L-cysteinyl-glycine-3C2,15N Sodium Salt
SMILES O[13C]([13C]([H])([15N](C([C@@H](NC(CC[C@H](N)C(O)=O)=O)CS)=O)[H])[H])=O

Specifications

Quantity 1 mg
Percent Purity >95%
Chemical Name or Material Glutathione (glycine-13C2,15N) Sodium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.