missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Peroxidase 1/GPX1 Antibody (C5-A10-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-32389
This item is not returnable.
View return policy
Description
Glutathione Peroxidase 1/GPX1 Monoclonal antibody specifically detects Glutathione Peroxidase 1/GPX1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Glutathione Peroxidase 1/GPX1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:1000 | |
| Cellular glutathione peroxidase, EC 1.11.1, EC 1.11.1.9, glutathione peroxidase 1, GPx-1, GSHPx-1, GSHPX1, MGC14399, MGC88245 | |
| Recombinant protein within Human Glutathione Peroxidase 1 aa 20-203/ 203. (Uniprot: P07203) | |
| 100 μL | |
| Apoptosis, Cancer, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 2876 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| C5-A10-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction