missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Peroxidase 4/GPX4 Mouse anti-Human, Mouse, Rat, Clone: LHM 2, Novus Biologicals™
Mouse Monoclonal Antibody
€ 519.00
Specifications
| Antigen | Glutathione Peroxidase 4/GPX4 |
|---|---|
| Clone | LHM 2 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
Glutathione Peroxidase 4/GPX4 Monoclonal specifically detects Glutathione Peroxidase 4/GPX4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Glutathione Peroxidase 4/GPX4 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| EC 1.11.1, EC 1.11.1.12, Glutathione peroxidase 4, glutathione peroxidase 4 (phospholipid hydroperoxidase), GPx-4, GSHPx-4, MCSP, PHGPxsnGPx, phospholipid hydroperoxidase, phospholipid hydroperoxide glutathione peroxidase, mitochondrial, snPHGPx, sperm nucleus glutathione peroxidase | |
| A 375P cells crude extract | |
| Primary | |
| Store at 4°C. |
| LHM 2 | |
| Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 2879 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title