missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Peroxidase 7 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-41103
This item is not returnable.
View return policy
Description
Glutathione Peroxidase 7 Polyclonal specifically detects Glutathione Peroxidase 7 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Glutathione Peroxidase 7 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CL683, EC 1.11.1, FLJ14777, glutathione peroxidase 6, glutathione peroxidase 7, GPX6EC 1.11.1.9, GPx-7, GSHPx-7, non-selenocysteine containing phospholipid hydroperoxide glutathione peroxidase, NPGPx | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/mL, Peptide ELISA Detection limit 1:32000 | |
| NP_056511.2 | |
| GPX7 | |
| Peptide with sequence CNQFGQQEPDSNK corresponding to internal region according to NP_056511.2. | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 2882 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction