missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Reductase Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97945AF594
This item is not returnable.
View return policy
Description
Glutathione Reductase Polyclonal specifically detects Glutathione Reductase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Glutathione Reductase | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 1.8.1, EC 1.8.1.7, GLUR, glutathione reductase, glutathione reductase, mitochondrial, GR, GRase, GRD1, MGC78522 | |
| Produced in rabbits immunized with E. coli-derived Human Glutathione Reductase fragment. | |
| 0.1 mL | |
| Cancer | |
| 2936 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction