missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Reductase Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99476MFV610
This item is not returnable.
View return policy
Description
Glutathione Reductase Polyclonal antibody specifically detects Glutathione Reductase in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| mFluor Violet 610 SE | |
| EC 1.8.1, EC 1.8.1.7, GLUR, glutathione reductase, glutathione reductase, mitochondrial, GR, GRase, GRD1, MGC78522 | |
| 0.1 mL | |
| 2936 | |
| IgG |
| 50mM Sodium Borate | |
| Antigen and protein A Affinity-purified | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction