missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione S-Transferase pi 1/GSTP1 Antibody (3F2C2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB110-60512-0.025ml
This item is not returnable.
View return policy
Description
Glutathione S-Transferase pi 1/GSTP1 Monoclonal specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Glutathione S-Transferase pi 1/GSTP1 | |
| Monoclonal | |
| Unconjugated | |
| P09211 | |
| GSTP1 | |
| Ni-NTA purified truncated recombinant GSTP1 expressed in E. Coli strain BL21 (DE3) | |
| Unpurified | |
| RUO | |
| Primary | |
| GSTP1 (3F2C2) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 3F2C2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI | |
| Mouse | |
| 23 kDa | |
| 0.025 mL | |
| Asthma, Cancer, Immunology | |
| 2950 | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction