missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione S-Transferase pi 1/GSTP1 Antibody (OTI4B6), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70857AF647
This item is not returnable.
View return policy
Description
Glutathione S-Transferase pi 1/GSTP1 Monoclonal specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Glutathione S-Transferase pi 1/GSTP1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GSTP1 | |
| Full length recombinant protein of human Glutathione S-Transferase pi 1/GSTP1 (NP_000843) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI4B6 | |
| Western Blot, Immunohistochemistry | |
| deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI | |
| Mouse | |
| Affinity Purified | |
| Asthma, Cancer, Immunology, Neuroscience | |
| 2950.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction