missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Synthetase Antibody (OTI1B8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70854F
This item is not returnable.
View return policy
Description
Glutathione Synthetase Monoclonal specifically detects Glutathione Synthetase in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| Glutathione Synthetase | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| GSS | |
| Full length recombinant protein of human Glutathione Synthetase (NP_000169) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| OTI1B8 | |
| Flow Cytometry, Immunohistochemistry | |
| EC 6.3.2.3, Glutathione synthase, glutathione synthetase, GSH synthetase, GSHS, GSH-S, MGC14098 | |
| Mouse | |
| Affinity Purified | |
| Neuroscience | |
| 2937.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction