missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLYATL1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98175AF405
This item is not returnable.
View return policy
Description
GLYATL1 Polyclonal antibody specifically detects GLYATL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GLYATL1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 92292 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| Acyl-CoA:glycine N-acyltransferase-like protein 1, EC 2.3.1.13, FLJ26507, FLJ34646, GATF-C, glycine N-acyltransferase-like protein 1, glycine-N-acyltransferase-like 1, GNAT, MGC15397, MGC15937 | |
| Produced in rabbits immunized with E. coli-derived Human GLYATL1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction