missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glycerol kinase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549801
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Glycerol kinase protein.
The product of this gene belongs to the FGGY kinase family of proteins and encodes glycerol kinase. Glycerol kinase is a key enzyme in the regulation of glycerol uptake and metabolism. It catalyzes the phosphorylation of glycerol by ATP, yielding ADP and glycerol-3-phosphate. Defects in this gene are the cause of glycerol kinase deficiency (GKD). Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Glycerol kinase | |
| Polyclonal | |
| Unconjugated | |
| GK | |
| ASTP; ATP:glycerol 3-phosphotransferase; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; D930012N15Rik; EC 2.7.1.30; GK; GK 3; GK1; GK3P; GKD; GKl; GKP3; GKTB; glycerokinase; Glycerokinase 3; glycerol kinase; Glycerolkinase; Gyk | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14933, 2710, 79223 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P32189, Q63060, Q64516 | |
| GK | |
| Synthesized peptide derived from C-terminal of human GLPK. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?