Learn More
Glyceryl Tritridecanoate, TRC
Product Code. 30404584
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404584 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30404584 Supplier Toronto Research Chemicals Supplier No. TRCG5982402.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Glyceryl Tritridecanoate
CAS 26536-12-9
Purity Grade Notes HPLC
Molecular Formula C42H80O6
Quantity 2.5 g
Synonym Tridecanoic Acid 1,2,3-Propanetriyl Ester, Glycerin Tritridecanoate, Glycerol tris(tridecanoate), Tritridecanoin, Tridecanoic Acid 1,1’,1’’-(1,2,3-Propanetriyl) Ester
InChI Formula InChI=1S/C42H80O6/c1-4-7-10-13-16-19-22-25-28-31-34-40(43)46-37-39(48-42(45)36-33-30-27-24-21-18-15-12-9-6-3)38-47-41(44)35-32-29-26-23-20-17-14-11-8-5-2/h39H,4-38H2,1-3H3
SMILES CCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCC)OC(=O)CCCCCCCCCCCC
IUPAC Name 2,3-di(tridecanoyloxy)propyl tridecanoate
Molecular Weight (g/mol) 681.08
Formula Weight 680.5955
Percent Purity >95