missing translation for 'onlineSavingsMsg'
Learn More

Glycine-13C, TRC

Product Code. 30405990
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405990 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405990 Supplier Toronto Research Chemicals Supplier No. TRCG615994250MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycine-13C
CAS 20220-62-6
Molecular Formula C13CH5NO2
Quantity 250 mg
Synonym 2-Aminoacetic Acid-13C, Aciport-13C, Aminoethanoic Acid-13C, Glicoamin-13C, Glycocoll-13C, Glycolixir-13C, Glycosthene-13C, Gyn-Hydralin-13C, NSC 25936-13C, NSC 2916-13C, NSC 54188-13C, Padil-13C
InChI Formula InChI=1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i1+1
SMILES OC([13CH2]N)=O
Molecular Weight (g/mol) 76.06
Formula Weight 76.0354
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.