missing translation for 'onlineSavingsMsg'
Learn More

Glycine-13C2,15N p-Toluenesulfonate, TRC

Product Code. 30408588
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408588 5 mg 5mg
30391612 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30408588 Supplier Toronto Research Chemicals Supplier No. TRCG6160175MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycine-13C2,15N p-Toluenesulfonate
CAS 2727323-57-9
Molecular Formula 13C2H515NO2 . C7H8O3S
Quantity 5 mg
InChI Formula InChI=1S/C7H8O3S.C2H5NO2/c1-6-2-4-7(5-3-6)11(8,9)10;3-1-2(4)5/h2-5H,1H3,(H,8,9,10);1,3H2,(H,4,5)/i;1+1,2+1,3+1
SMILES O[13C]([13CH2][15NH2])=O.CC1=CC=C(C=C1)S(=O)(O)=O
Molecular Weight (g/mol) 250.25
Formula Weight 250.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.