Learn More
Glycine N-Methyltransferase/GNMT Antibody (5D1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59452PE
This item is not returnable.
View return policy
Description
Glycine N-Methyltransferase/GNMT Monoclonal antibody specifically detects Glycine N-Methyltransferase/GNMT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Glycine N-Methyltransferase/GNMT | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 5D1 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| EC 2.1.1.20, glycine N-methyltransferase | |
| Recombinant human Glycine N-Methyltransferase/GNMT (1-295aa) purified from E. coli | |
| 0.1 mL | |
| Vision | |
| 27232 | |
| Store at 4C in the dark. | |
| IgG2a κ |