missing translation for 'onlineSavingsMsg'
Learn More

Glycocholic acid hydrate, Thermo Scientific™

Product Code. p-7237787 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17211316

Brand: Thermo Scientific Acros 466790010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192657-83-2
Molecular Formula C26H43NO6
Molecular Weight (g/mol) 465.63
MDL Number MFCD06408004
InChI Key RFDAIACWWDREDC-MTSIQNSJNA-N
IUPAC Name 2-(4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)acetic acid hydrate
SMILES [H][C@@]1(CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC(=O)NCC(O)=O

Specifications

Alpha Vector Glycocholicacid
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 1 g
Solubility Information (50 mg/ml Ethanol) Clear colorless to light yellow solution
Formula Weight 483.64
Physical Form Powder
Chemical Name or Material Glycocholic acid hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.