missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glycogen Synthase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29755
This item is not returnable.
View return policy
Description
Glycogen Synthase Polyclonal antibody specifically detects Glycogen Synthase in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Glycogen Synthase | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 2.4.1.11, glycogen [starch] synthase, muscle, glycogen synthase 1 (muscle), GYSGSY | |
| Between 625 and 675 | |
| 100 μg | |
| Phospho Specific | |
| 2997 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction