missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycogen Synthase [p Ser641] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33559-100ul
This item is not returnable.
View return policy
Description
Glycogen Synthase Monoclonal antibody specifically detects Glycogen Synthase in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Glycogen Synthase | |
| Monoclonal | |
| Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL | |
| EC 2.4.1.11, glycogen [starch] synthase, muscle, glycogen synthase 1 (muscle), GYSGSY | |
| A synthetic phosphorylated peptide around S641 of human Glycogen Synthase (GYS1) (P13807).,, Sequence:, PASVP | |
| 100 μL | |
| Phospho Specific | |
| 2997 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction