missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycogenin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | Glycogenin 1 |
|---|---|
| Dilution | Western Blot 1:200 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231284
|
Novus Biologicals
NBP3-35107-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230501
|
Novus Biologicals
NBP3-35107-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycogenin 1 Polyclonal antibody specifically detects Glycogenin 1 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Glycogenin 1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 2992 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.4.1.186, glycogenin, glycogenin 1, glycogenin-1, GN1, GN-1, GSD15, GYG | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 234-333 of human Glycogenin 1 (NP_001171649.1).,, Sequence:, EAHDPNMTHPEFLILWWNIFTTNVLPLLQQFGLVKDTCSYVNVEDVSGAISHLSLGEIPAMAQPFVSSEERKERWEQGQADYMGADSFDNIKRKLDTYLQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title