missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | Glycophorin C |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 μL/mg of lysate, Immunohistochemistry-Paraffin 1:1000-1:10000 |
| Applications | Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666682
|
Novus Biologicals
NBP2-97086-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688974
|
Novus Biologicals
NBP2-97086-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycophorin C Polyclonal antibody specifically detects Glycophorin C in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| Glycophorin C | |
| Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 2995 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 μL/mg of lysate, Immunohistochemistry-Paraffin 1:1000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2', Sialoglycoprotein D | |
| Produced in rabbits immunized with purified, recombinant Human Glycophorin C (Accession#: NP_002092.1; Met1-Met57) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title