missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin C Antibody (SR1653), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Glycophorin C |
|---|---|
| Clone | SR1653 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653444
|
Novus Biologicals
NBP3-21888-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654044
|
Novus Biologicals
NBP3-21888-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycophorin C Monoclonal antibody specifically detects Glycophorin C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| Glycophorin C | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2', Sialoglycoprotein D | |
| A synthesized peptide derived from human Glycophorin C (Uniprot #: P04921) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1653 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2995 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title