missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLYCTK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 183.00 - € 304.50
Specifications
| Antigen | GLYCTK |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683797
|
Novus Biologicals
NBP3-12587-100ul |
100 μg |
€ 322.00 € 304.50 / 100µL Save € 17.50 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665167
|
Novus Biologicals
NBP3-12587-50ul |
50 μg |
€ 183.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLYCTK Polyclonal antibody specifically detects GLYCTK in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| GLYCTK | |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| PBS, pH7.0 | |
| 132158 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.1.31, glycerate kinase, GLYCTK1, HBeAg-binding protein 2, HBeAg-binding protein 4, HBeAgBP4A, HBEBP2, HBEBP4HBeAg binding protein 4 | |
| Produced in rabbits immunized with E. coli-derived Human GLYCTK fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title