Learn More
Glycyl-D-phenylalanine, TRC
Specifications
Specifications
| Chemical Name or Material | Glycyl-D-phenylalanine |
| CAS | 34258-14-5 |
| Molecular Formula | C11 H14 N2 O3 |
| Quantity | 25 mg |
| Synonym | Glycyl-D-phenylalanine, (2R)-2-[(2-Azaniumylacetyl)amino]-3-phenylpropanoate, (R)-2-(2-Aminoacetylamino)-3-phenylpropionic Acid, Gly-D-Phe, NSC 522450 |
| InChI Formula | InChI=1S/C11H14N2O3/c12-7-10(14)13-9(11(15)16)6-8-4-2-1-3-5-8/h1-5,9H,6-7,12H2,(H,13,14)(H,15,16)/t9-/m1/s1 |
| SMILES | NCC(=O)N[C@H](Cc1ccccc1)C(=O)O |
| IUPAC Name | (2R)-2-[(2-aminoacetyl)amino]-3-phenyl-propanoic acid |
| Molecular Weight (g/mol) | 222.24 |
| Formula Weight | 222.1004 |