Learn More
Thermo Scientific Chemicals Glycylglycine, 99+%
Catalog Number p-139252
Click to view available options
Quantity:
25 g
100 g
1 kg
556-50-3
C4H8N2O3
132.12
MFCD00008130
YMAWOPBAYDPSLA-UHFFFAOYSA-N
glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
11163
CHEBI:17201
2-[(2-aminoacetyl)amino]acetic acid
C(C(=O)NCC(=O)O)N
556-50-3
C4H8N2O3
132.12
MFCD00008130
YMAWOPBAYDPSLA-UHFFFAOYSA-N
glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
11163
CHEBI:17201
2-[(2-aminoacetyl)amino]acetic acid
C(C(=O)NCC(=O)O)N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
TRUSTED_SUSTAINABILITY
Melting Point 262.0°C to 264.0°C
Absorbance (10% aq. soln.) (1 cm cell), 0.01 max. at 405nm
pH 5.5 to 6 (5% aq. soln.)
Color White
Chloride 100ppm max
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula H2NCH2CONHCH2CO2H
Beilstein 04, 371
Loss on Drying 0.5% max. (105°C, 4 hrs)
Merck Index 15, 4539
Packaging Glass Bottle
Solubility Information Solubility in water: 132g/L (20°C). Other solubilities: slightly soluble in ethanol, practically insoluble in ether
Formula Weight 132.12
Percent Purity ≥99%
Sulfated Ash 0.1% max.
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Glycylglycine, 99+%