missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxalase I Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98271FR
This item is not returnable.
View return policy
Description
Glyoxalase I Polyclonal specifically detects Glyoxalase I in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Glyoxalase I | |
| Polyclonal | |
| Western Blot, ELISA | |
| Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
| Produced in rabbits immunized with purified, recombinant Glyoxalase I (NP_079650.3; Ala 2-Ile 184). | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 2739 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction