missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Glypican 3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16630334
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16630334 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16630334 Supplier Invitrogen™ Supplier No. PA5102528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPC3.

GPC3 is a cell surface proteoglycan that bears heparan sulfate. This protein may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol (GPI) linkage. These proteins may play a role in the control of cell division, growth regulation, and tumor predisposition. Deletion mutations in GPC3 are the cause of Simpson-Golabi-Behmel syndrome (SGBS), also known as Simpson dysmorphia syndrome (SDYS). SGBS is a condition characterized by pre- and postnatal overgrowth (gigantism) with visceral and skeletal anomalies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPC3
Gene Accession No. P13265, P51654, Q8CFZ4
Gene Alias defective in Simpson-Golabi-Behmel overgrowth syndrome; DGSX; glypican 3; glypican proteoglycan 3; glypican-3; Glypican-3 alpha subunit; Glypican-3 beta subunit; Gpc3; GTR22; GTR2-2; heparan sulphate proteoglycan; intestinal protein OCI-5; MXR7; OCI5; OCI-5; proteoglycan GPC3; SDYS; secreted glypican-3; SGB; SGBS; SGBS1
Gene Symbols GPC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPC3(Accession P51654), corresponding to amino acid residues R39-M89.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14734, 25236, 2719
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.