missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican 4 Antibody (961609) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9195X
This item is not returnable.
View return policy
Description
Glypican 4 Monoclonal antibody specifically detects Glypican 4 in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF
Specifications
| Glypican 4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, CyTOF | |
| 961609 | |
| Flow Cytometry, ELISA, CyTOF-ready | |
| glypican 4, glypican proteoglycan 4, glypican-4, K-glypicandJ900E8.1 (glypican 4) | |
| Chinese hamster ovary cell line CHO-derived recombinant human Glypican 4, Met1-Ser529, Accession # O75487 | |
| 0.1 mL | |
| Cancer, Stem Cells | |
| 2239 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction