missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glypican 4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597801
This item is not returnable.
View return policy
Description
Glypican 4 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. The GPC4 gene is adjacent to the 3' end of GPC3 and may also play a role in Simpson-Golabi-Behmel syndrome.
Specifications
| Glypican 4 | |
| Polyclonal | |
| Unconjugated | |
| GPC4 | |
| 9530073D23Rik; AF311610; AI385680; AI661372; dJ900E8.1 (glypican 4); glypican 4; glypican proteoglycan 4; glypican-4; Gpc4; K-glypican; Secreted glypican-4; UNQ474/PRO937 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 14735, 2239 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O75487, P51655 | |
| GPC4 | |
| Recombinant Human Glypican-4 protein (19-210AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction