Learn More
Invitrogen™ GlyT2 Recombinant Rabbit Monoclonal Antibody (23GB4145), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552662
 View more versions of this product
Product Code. 30089671
  • € 446.00 / 100µL
Description
This gene encodes a sodium- and chloride-dependent glycine neurotransmitter transporter. This integral membrane glycoprotein is responsible for the clearance of extracellular glycine during glycine-mediated neurotransmission.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GlyT2
Recombinant Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P58295, Q761V0, Q9Y345
SLC6A5
A synthesized peptide derived from human Glyt2 (301-336AA).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB4145
Unconjugated
SLC6A5
glycine transporter 2; GLYT2; glyT-2; HKPX3; NET1; prestin; si:dkey-7n10.12; slc6a5; Sodium- and chloride-dependent glycine transporter 2; solute carrier family 6 (neurotransmitter transporter), member 5; solute carrier family 6 (neurotransmitter transporter, glycine), member 5; solute carrier family 6 member 5
Rabbit
Affinity chromatography
RUO
104245, 171148, 9152
-20°C