Learn More
GM-CSF Antibody (83308), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB5181D
Product Code. 30061543
  • € 896.00 / 0.50mL
Description
GM-CSF Monoclonal antibody specifically detects GM-CSF in Rat samples. It is validated for Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GM-CSF
Monoclonal
DyLight 350
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Rat
Purified
Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF
83308
Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
E. coli-derived recombinant rat GM-CSF, Ala1-Lys127, Accession # P48750
0.5 mL
Biologically Active Proteins, Cytokine Research, Immunology
1437
Store at 4C in the dark.
IgG2b