missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM-CSFR alpha Antibody (4H1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43361PCP
This item is not returnable.
View return policy
Description
GM-CSFR alpha Monoclonal specifically detects GM-CSFR alpha in Human samples. It is validated for Flow Cytometry.
Specifications
| GM-CSFR alpha | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| CSF2RA | |
| The immunogen for this antibody was CD116. | |
| 0.1 mL | |
| Cancer | |
| 1438 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| 4H1 | |
| Flow Cytometry | |
| CD116, CD116 antigen, CDw116, colony stimulating factor 2 receptor alpha subunit, colony stimulating factor 2 receptor, alpha, low-affinity(granulocyte-macrophage), CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF receptor alpha subunit, GMCSFR, GM-CSF-R-alpha, GMCSFR-alpha, GMR, GMR-alpha, granulocyte-macrophage colony-stimulating factor receptor alpha chain, granulocyte-macrophage colony-stimulating factor receptor subunit alpha, MGC3848, MGC4838 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction