missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM130/GOLGA2 Antibody (2059D), Alexa Fluor™ 488, R&D Systems™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB8199AF488
This item is not returnable.
View return policy
Description
GM130/GOLGA2 Monoclonal specifically detects GM130/GOLGA2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| GM130/GOLGA2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GOLGA2 | |
| Partial recombinant human GOLG-A2 protein (amino acids 528-606) [UniProt Q08379] | |
| 100 μL | |
| Cellular Markers, Golgi Apparatus Markers | |
| 2801 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 2059D | |
| Western Blot, Immunohistochemistry | |
| 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction