missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM130/GOLGA2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-53420APC
This item is not returnable.
View return policy
Description
GM130/GOLGA2 Polyclonal antibody specifically detects GM130/GOLGA2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| GM130/GOLGA2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| APC | |
| 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
| Partial recombinant human GM130/GOLGA2 protein (amino acids 528-606). [UniPro Q08379] | |
| 0.1 mL | |
| Cellular Markers, Golgi Apparatus Markers | |
| 2801 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction