Learn More
GM130/GOLGA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP1-89756
 View more versions of this product
Product Code. 18787503
  • € 557.00 / 0.10mL
Description
GM130/GOLGA2 Polyclonal specifically detects GM130/GOLGA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunofluorescence
Affinity Purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein
0.1 mL