missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM130/GOLGA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 369.00 - € 529.00
Specifications
| Antigen | GM130/GOLGA2 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18435241
|
Novus Biologicals
NBP1-89757-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18285656
|
Novus Biologicals
NBP1-89757 |
0.1 mL |
€ 529.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GM130/GOLGA2 Polyclonal specifically detects GM130/GOLGA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GM130/GOLGA2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
| GOLGA2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Golgi Apparatus Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 2801 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DTPKDNAATLQPSDDTVLPGGVPSPGASLTSMAASQNHDADNVPNLMDETKTFSSTESLRQLSQQLNGLVCESATCVNGEGPASSANLK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title