missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GM130 Recombinant Rabbit Monoclonal Antibody (2L3P6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535107
This item is not returnable.
View return policy
Description
Immunogen sequence: MWPQPRLPPR PAMSEETRQS KLAAAKKKLR EYQQRNSPGV PTGAKKKKKI KNGSNPETTT SGGCHSPEDT PKDNAATLQP SDDTVLPGGV PSPGASLTSM.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined.
Specifications
| GM130 | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.09% sodium azide, pH 7.3 | |
| Q08379, Q921M4 | |
| Golga2 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2L3P6 | |
| Unconjugated | |
| Golga2 | |
| 130 kDa cis-Golgi matrix protein; AW555139; cis-Golgi matrix protein GM130; Gm130; GM130 autoantigen; GOLGA2; golgi autoantigen, golgin subfamily a, 2; Golgi matrix protein GM130; golgin A2; Golgin subfamily A member 2; Golgin subfamily A member 2; LOW QUALITY PROTEIN: golgin subfamily A member 2; golgin-95; mKIAA4150; RP11-395P17.5; SY11 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2801, 99412 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction