missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM2A Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00106APC
This item is not returnable.
View return policy
Description
GM2A Polyclonal antibody specifically detects GM2A in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| GM2A | |
| Polyclonal | |
| Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| Cerebroside sulfate activator protein, ganglioside GM2 activator, GM2 ganglioside activator, GM2-AP, SAP-3GM2 ganglioside activator protein, Shingolipid activator protein 3, sphingolipid activator protein 3 | |
| Produced in rabbits immunized with purified, recombinant Human GM2A (Accession#: AAA35907.1; Met1-Ile193) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 2760 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction